Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- MSTO1
- Entrez Gene ID
-
55154
- Gene Name
- misato 1, mitochondrial distribution and morphology regulator
- Gene Aliases
-
LST005, MST
- Chromosome Location
-
Chr.1: 155610170 - 155614967 on Build GRCh38
- Gene Symbol
- MSTO2P
- Entrez Gene ID
-
100129405
- Gene Name
- misato family member 2, pseudogene
- Gene Aliases
-
MSTO2
- Chromosome Location
-
Chr.1: 155745768 - 155750688 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_024117.2 |
|
6 - 7 |
685 |
82 |
GenBank mRNA |
AK293928.1 |
- |
6 - 7 |
621 |
82 |
AK295146.1 |
- |
6 - 7 |
685 |
82 |
BC099909.1 |
- |
6 - 7 |
635 |
82 |
View Full Product Details