Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ANKRD24
Entrez Gene ID
170961
Gene Name
ankyrin repeat domain 24
Gene Aliases
-
Chromosome Location
Chr.19: 4182618 - 4224814 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BI914435.1 - 5 - 6 437 126

View Full Product Details