The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_017027016.1 | XP_016882505.1 | 7 - 8 | 975 | 73 |
GenBank mRNA | AL365405.1 | - | 2 - 3 | 383 | 73 |