The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000698.4 | NP_000689.1 | 12 - 13 | 1782 | 154 |
NM_001256153.2 | NP_001243082.1 | 12 - 13 | 1686 | 154 | |
NM_001320861.1 | NP_001307790.1 | 12 - 13 | 1695 | 154 | |
NM_001320862.1 | NP_001307791.1 | 12 - 13 | 1820 | 154 | |
XM_017016012.1 | XP_016871501.1 | 11 - 12 | 1657 | 154 | |
GenBank mRNA | AB208946.1 | - | 12 - 13 | 4452 | 154 |
AK315223.1 | - | 12 - 13 | 1636 | 154 | |
BC034464.1 | - | 12 - 13 | 1721 | 154 | |
BC040450.1 | - | 12 - 13 | 1711 | 154 | |
BC130332.1 | - | 12 - 13 | 1735 | 154 | |
BC132677.1 | - | 12 - 13 | 1735 | 154 | |
HM592260.1 | - | 11 - 12 | 1867 | 154 | |
HM592261.1 | - | 12 - 13 | 1572 | 154 | |
J03571.1 | - | 12 - 13 | 1702 | 154 | |
J03600.1 | - | 12 - 13 | 1712 | 154 | |
JQ728550.1 | - | 11 - 12 | 1588 | 154 |