The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000698.4 | NP_000689.1 | 6 - 7 | 949 | 61 |
NM_001256153.2 | NP_001243082.1 | 6 - 7 | 949 | 61 | |
NM_001256154.2 | NP_001243083.1 | 6 - 7 | 949 | 61 | |
NM_001320861.1 | NP_001307790.1 | 6 - 7 | 949 | 61 | |
NM_001320862.1 | NP_001307791.1 | 6 - 7 | 987 | 61 | |
XM_017016012.1 | XP_016871501.1 | 5 - 6 | 824 | 61 | |
GenBank mRNA | AB208946.1 | - | 7 - 8 | 3220 | 61 |
AK225603.1 | - | 6 - 7 | 902 | 61 | |
AK315223.1 | - | 6 - 7 | 899 | 61 | |
BC034464.1 | - | 6 - 7 | 888 | 61 | |
BC040450.1 | - | 6 - 7 | 878 | 61 | |
BC130332.1 | - | 6 - 7 | 902 | 61 | |
BC132677.1 | - | 6 - 7 | 902 | 61 | |
BC143985.1 | - | 6 - 7 | 902 | 61 | |
HM592258.1 | - | 6 - 7 | 835 | 61 | |
HM592259.1 | - | 6 - 7 | 835 | 61 | |
HM592260.1 | - | 6 - 7 | 835 | 61 | |
HM592261.1 | - | 6 - 7 | 835 | 61 | |
J03571.1 | - | 6 - 7 | 869 | 61 | |
J03600.1 | - | 6 - 7 | 879 | 61 | |
JQ728550.1 | - | 5 - 6 | 755 | 61 |