The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001191056.3 | NP_001177985.1 | 7 - 8 | 1215 | 91 |
NM_001191057.3 | NP_001177986.1 | 7 - 8 | 1215 | 91 | |
NM_001191058.3 | NP_001177987.2 | 8 - 9 | 1595 | 91 | |
NM_001191059.3 | NP_001177988.1 | 8 - 9 | 1206 | 91 | |
NM_001322055.1 | NP_001308984.1 | 8 - 9 | 1441 | 91 | |
NM_001322056.1 | NP_001308985.1 | 8 - 9 | 1206 | 91 | |
NM_001322057.1 | NP_001308986.1 | 8 - 9 | 1441 | 91 | |
NM_001322058.1 | NP_001308987.1 | 8 - 9 | 1595 | 91 | |
NM_001322059.1 | NP_001308988.1 | 8 - 9 | 1244 | 91 | |
NM_005020.4 | NP_005011.1 | 8 - 9 | 979 | 91 | |
XM_017012264.1 | XP_016867753.1 | 8 - 9 | 1046 | 91 | |
XM_017012265.1 | XP_016867754.1 | 8 - 9 | 897 | 91 | |
XM_017012266.1 | XP_016867755.1 | 8 - 9 | 1046 | 91 | |
XM_017012267.1 | XP_016867756.1 | 8 - 9 | 951 | 91 | |
GenBank mRNA | AK056170.1 | - | 7 - 8 | 1202 | 91 |
AK091734.1 | - | 8 - 9 | 1520 | 91 | |
AK094180.1 | - | 8 - 9 | 1138 | 91 | |
BC022479.1 | - | 7 - 8 | 1216 | 91 | |
BC022525.1 | - | 7 - 8 | 1216 | 91 | |
U40371.1 | - | 8 - 9 | 922 | 91 | |
U40372.1 | - | 3 - 4 | 312 | 91 |