The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015056.2 | NP_055871.1 | 6 - 7 | 663 | 146 |
GenBank mRNA | AK056263.1 | - | 6 - 7 | 626 | 146 |
BC014005.2 | - | 6 - 7 | 635 | 146 | |
BC028386.2 | - | 6 - 7 | 668 | 146 | |
D80001.1 | - | 6 - 7 | 561 | 146 |