Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ATXN7L1
Entrez Gene ID
222255
Gene Name
ataxin 7 like 1
Gene Aliases
ATXN7L4
Chromosome Location
Chr.7: 105604774 - 105877232 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006715900.2 XP_006715963.1 10 - 11 1628 118
XM_006715901.2 XP_006715964.1 10 - 11 1628 118
XM_006715902.3 XP_006715965.1 10 - 11 1628 118
XM_006715903.3 XP_006715966.1 10 - 11 1628 118
XM_006715904.3 XP_006715967.1 6 - 7 1894 118
XM_011515935.1 XP_011514237.1 10 - 11 1628 118
XM_011515936.2 XP_011514238.1 8 - 9 2124 118
XM_011515938.2 XP_011514240.1 7 - 8 1975 118
XM_011515939.1 XP_011514241.1 10 - 11 1628 118
XM_011515941.2 XP_011514243.1 8 - 9 2090 118
XM_011515942.2 XP_011514244.1 7 - 8 1624 118
XM_011515943.2 XP_011514245.1 7 - 8 1971 118
XM_011515944.1 XP_011514246.1 7 - 8 1206 118
XM_011515945.1 XP_011514247.1 8 - 9 1389 118
XM_011515946.1 XP_011514248.1 6 - 7 1123 118
XM_011515947.1 XP_011514249.1 6 - 7 1158 118
XM_017011858.1 XP_016867347.1 7 - 8 3111 118
XM_017011860.1 XP_016867349.1 8 - 9 1377 118
XM_017011861.1 XP_016867350.1 6 - 7 1559 118
GenBank mRNA BC003517.1 - 2 - 3 315 118

View Full Product Details