Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ATXN7L1
Entrez Gene ID
222255
Gene Name
ataxin 7 like 1
Gene Aliases
ATXN7L4
Chromosome Location
Chr.7: 105604774 - 105877232 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020725.1 NP_065776.1 4 - 5 612 85
NM_138495.1 NP_612504.1 2 - 3 341 85
XM_005250220.3 XP_005250277.1 5 - 6 695 85
XM_006715900.2 XP_006715963.1 5 - 6 695 85
XM_006715901.2 XP_006715964.1 5 - 6 695 85
XM_006715902.3 XP_006715965.1 5 - 6 695 85
XM_006715903.3 XP_006715966.1 5 - 6 695 85
XM_006715904.3 XP_006715967.1 2 - 3 1044 85
XM_011515935.1 XP_011514237.1 5 - 6 695 85
XM_011515936.2 XP_011514238.1 3 - 4 1191 85
XM_011515937.2 XP_011514239.1 5 - 6 695 85
XM_011515938.2 XP_011514240.1 2 - 3 1042 85
XM_011515939.1 XP_011514241.1 5 - 6 695 85
XM_011515940.1 XP_011514242.1 5 - 6 695 85
XM_011515941.2 XP_011514243.1 3 - 4 1157 85
XM_011515942.2 XP_011514244.1 2 - 3 691 85
XM_011515943.2 XP_011514245.1 2 - 3 1038 85
XM_011515944.1 XP_011514246.1 2 - 3 273 85
XM_011515946.1 XP_011514248.1 2 - 3 273 85
XM_011515947.1 XP_011514249.1 2 - 3 308 85
XM_011515950.1 XP_011514252.1 5 - 6 695 85
XM_017011858.1 XP_016867347.1 2 - 3 2178 85
XM_017011859.1 XP_016867348.1 2 - 3 1034 85
XM_017011860.1 XP_016867349.1 4 - 5 527 85
XM_017011861.1 XP_016867350.1 2 - 3 709 85
GenBank mRNA AB033044.1 - 4 - 5 625 85
AK300338.1 - 2 - 3 341 85
AK302013.1 - 2 - 3 308 85
BQ227169.1 - 2 - 3 385 85
BQ436521.1 - 3 - 4 437 85

View Full Product Details