The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001300959.1 | NP_001287888.1 | 6 - 7 | 852 | 118 |
NM_031449.3 | NP_113637.3 | 6 - 7 | 943 | 118 | |
NM_174929.2 | NP_777589.2 | 5 - 6 | 882 | 118 | |
XM_005249866.2 | XP_005249923.1 | 6 - 7 | 967 | 118 | |
XM_005249867.4 | XP_005249924.1 | 6 - 7 | 969 | 118 | |
XM_005249868.4 | XP_005249925.1 | 6 - 7 | 1987 | 118 | |
XM_005249869.2 | XP_005249926.1 | 6 - 7 | 967 | 118 | |
XM_005249870.2 | XP_005249927.1 | 6 - 7 | 967 | 118 | |
XM_005249871.2 | XP_005249928.1 | 6 - 7 | 871 | 118 | |
XM_005249872.2 | XP_005249929.1 | 6 - 7 | 967 | 118 | |
XM_005249873.2 | XP_005249930.1 | 6 - 7 | 871 | 118 | |
XM_017012674.1 | XP_016868163.1 | 6 - 7 | 1048 | 118 | |
GenBank mRNA | AB067473.1 | - | 1 - 2 | 105 | 118 |
AF357908.1 | - | 5 - 6 | 820 | 118 | |
AK090415.1 | - | 6 - 7 | 943 | 118 | |
AY426594.1 | - | 5 - 6 | 844 | 118 | |
BC016968.1 | - | 5 - 6 | 865 | 118 | |
BC110435.1 | - | 6 - 7 | 837 | 118 |