Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TESC
Entrez Gene ID
54997
Gene Name
tescalcin
Gene Aliases
CHP3, TSC
Chromosome Location
Chr.12: 117038923 - 117099490 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001168325.1 NP_001161797.1 4 - 5 501 72
NM_017899.3 NP_060369.3 5 - 6 582 72
XM_011538503.1 XP_011536805.1 5 - 6 397 72
XM_017019532.1 XP_016875021.1 5 - 6 626 72
GenBank mRNA AF443207.1 - 5 - 6 539 72
AI910454.1 - 3 - 4 72
BC015221.1 - 5 - 6 539 72
BE670015.1 - 3 - 4 72
BQ949772.1 - 4 - 5 455 72

View Full Product Details