Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
UBE3B
Entrez Gene ID
89910
Gene Name
ubiquitin protein ligase E3B
Gene Aliases
BPIDS, KOS
Chromosome Location
Chr.12: 109477410 - 109543628 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_130466.3 NP_569733.2 26 - 27 3537 121
NM_183415.2 NP_904324.1 26 - 27 3192 121
XM_005253987.2 XP_005254044.1 26 - 27 3417 121
XM_006719681.3 XP_006719744.1 26 - 27 3750 121
XM_006719682.2 XP_006719745.1 26 - 27 3750 121
XM_011538959.2 XP_011537261.1 25 - 26 3299 121
XM_011538961.1 XP_011537263.1 26 - 27 3750 121
XM_017020195.1 XP_016875684.1 26 - 27 3960 121
GenBank mRNA AF130072.1 - 24 - 25 2929 121
AF251046.2 - 26 - 27 3507 121
AK127227.1 - 11 - 12 2324 121
AK301036.1 - 2 - 3 256 121
AK304678.1 - 2 - 3 202 121
AL096740.1 - 27 - 28 3610 121
BC020348.1 - 11 - 12 1231 121
BC032301.1 - 26 - 27 3170 121
BC141880.1 - 26 - 27 3415 121
BC144684.1 - 26 - 27 3513 121
BC150637.1 - 26 - 27 3168 121
BX537403.1 - 26 - 27 3182 121

View Full Product Details