The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001300801.1 | NP_001287730.1 | 6 - 7 | 1148 | 86 |
NM_020845.2 | NP_065896.1 | 6 - 7 | 1087 | 86 | |
XM_011538590.2 | XP_011536892.1 | 6 - 7 | 1112 | 86 | |
GenBank mRNA | AB040890.2 | - | 7 - 8 | 1169 | 86 |
AF334585.1 | - | 5 - 6 | 1026 | 86 | |
BC141813.1 | - | 6 - 7 | 1188 | 86 |