The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001923.4 | NP_001914.3 | 23 - 24 | 3179 | 69 |
GenBank mRNA | AJ002955.1 | - | 23 - 24 | 3044 | 69 |
AK294341.1 | - | 7 - 8 | 953 | 69 | |
AK294493.1 | - | 25 - 26 | 3308 | 69 | |
AK303032.1 | - | 17 - 18 | 2061 | 69 | |
AK312436.1 | - | 23 - 24 | 3001 | 69 | |
BC011686.2 | - | 23 - 24 | 3008 | 69 | |
BC032080.1 | - | 9 - 10 | 1108 | 69 | |
BC050530.1 | - | 23 - 24 | 3072 | 69 | |
BC051764.2 | - | 23 - 24 | 3062 | 69 | |
L40326.1 | - | 23 - 24 | 2973 | 69 | |
U18299.1 | - | 23 - 24 | 3035 | 69 | |
U32986.1 | - | 23 - 24 | 3044 | 69 |