The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001923.4 | NP_001914.3 | 3 - 4 | 572 | 96 |
GenBank mRNA | AK294493.1 | - | 5 - 6 | 701 | 96 |
AK312436.1 | - | 3 - 4 | 394 | 96 | |
AL831958.1 | - | 3 - 4 | 554 | 96 | |
BC011686.2 | - | 3 - 4 | 401 | 96 | |
BC021044.1 | - | 3 - 4 | 389 | 96 | |
BC050530.1 | - | 3 - 4 | 465 | 96 | |
BC051764.2 | - | 3 - 4 | 455 | 96 | |
DC334780.1 | - | 4 - 5 | 438 | 96 | |
L40326.1 | - | 3 - 4 | 366 | 96 | |
U18299.1 | - | 3 - 4 | 428 | 96 | |
U32986.1 | - | 3 - 4 | 437 | 96 |