Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC2A2
Entrez Gene ID
6514
Gene Name
solute carrier family 2 member 2
Gene Aliases
GLUT2
Chromosome Location
Chr.3: 170996341 - 171026979 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000340.1 NP_000331.1 7 - 8 1272 97
NM_001278658.1 NP_001265587.1 6 - 7 1009 97
NM_001278659.1 NP_001265588.1 6 - 7 1147 97
XM_011513087.2 XP_011511389.1 8 - 9 1019 97
XM_011513089.2 XP_011511391.1 4 - 5 560 97
GenBank mRNA AK290846.1 - 7 - 8 1015 97
AK292741.1 - 7 - 8 1015 97
AK298418.1 - 6 - 7 773 97
AK313622.1 - 7 - 8 1013 97
BC060041.1 - 6 - 7 892 97
J03810.1 - 7 - 8 1001 97

View Full Product Details