The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001606.4 | NP_001597.2 | 32 - 33 | 5339 | 70 |
NM_212533.2 | NP_997698.1 | 32 - 33 | 5331 | 70 | |
XM_006716996.3 | XP_006717059.1 | 31 - 32 | 5273 | 70 | |
GenBank mRNA | AB028985.2 | - | 18 - 19 | 3196 | 70 |
AB177854.1 | - | 19 - 20 | 3240 | 70 | |
AF178941.1 | - | 31 - 32 | 5241 | 70 | |
AF327657.1 | - | 31 - 32 | 5197 | 70 | |
AL833967.1 | - | 19 - 20 | 3279 | 70 | |
BC008755.1 | - | 3 - 4 | 486 | 70 | |
BC144588.1 | - | 31 - 32 | 5321 | 70 | |
BF983101.1 | - | 4 - 5 | 70 |