The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001606.4 | NP_001597.2 | 41 - 42 | 6498 | 144 |
NM_212533.2 | NP_997698.1 | 41 - 42 | 6490 | 144 | |
XM_006716996.3 | XP_006717059.1 | 40 - 41 | 6432 | 144 | |
GenBank mRNA | AB028985.2 | - | 27 - 28 | 4355 | 144 |
AB177854.1 | - | 27 - 28 | 4647 | 144 | |
AF178941.1 | - | 40 - 41 | 6400 | 144 | |
AF327657.1 | - | 40 - 41 | 6356 | 144 | |
AK295489.1 | - | 7 - 8 | 966 | 144 | |
AL162060.1 | - | 1 - 2 | 163 | 144 | |
BC008755.1 | - | 12 - 13 | 1645 | 144 | |
BC144588.1 | - | 39 - 40 | 6560 | 144 |