The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_144718.3 | NP_653319.1 | 3 - 4 | 443 | 89 |
XM_011512461.2 | XP_011510763.1 | 3 - 4 | 200 | 89 | |
XM_017005775.1 | XP_016861264.1 | 3 - 4 | 200 | 89 | |
XM_017005776.1 | XP_016861265.1 | 3 - 4 | 337 | 89 | |
XM_017005777.1 | XP_016861266.1 | 3 - 4 | 260 | 89 | |
XM_017005778.1 | XP_016861267.1 | 3 - 4 | 444 | 89 | |
GenBank mRNA | AK057966.1 | - | 3 - 4 | 224 | 89 |
AK296160.1 | - | 3 - 4 | 337 | 89 | |
AL832148.1 | - | 3 - 4 | 412 | 89 | |
AL832327.2 | - | 3 - 4 | 226 | 89 | |
AY099107.1 | - | 3 - 4 | 363 | 89 | |
BC036951.1 | - | 3 - 4 | 384 | 89 | |
BP278430.1 | - | 3 - 4 | 443 | 89 |