Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DCAF6
Entrez Gene ID
55827
Gene Name
DDB1 and CUL4 associated factor 6
Gene Aliases
1200006M05Rik, ARCAP, IQWD1, MSTP055, NRIP, PC326
Chromosome Location
Chr.1: 167936247 - 168075843 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017977.2 NP_001017977.1 16 - 17 2673 147
NM_001198956.1 NP_001185885.1 19 - 20 2946 147
NM_001198957.1 NP_001185886.1 18 - 19 2853 147
NM_018442.3 NP_060912.2 17 - 18 2733 147
XM_005245331.4 XP_005245388.1 18 - 19 2845 147
XM_005245332.4 XP_005245389.1 18 - 19 2716 147
XM_005245333.4 XP_005245390.1 17 - 18 2656 147
XM_017001779.1 XP_016857268.1 19 - 20 2759 147
XM_017001780.1 XP_016857269.1 17 - 18 2482 147
XM_017001781.1 XP_016857270.1 16 - 17 2472 147
GenBank mRNA AF116725.1 - 19 - 20 2687 147
AF124434.1 - 15 - 16 2164 147
AF150734.1 - 14 - 15 2033 147
AK226181.1 - 16 - 17 2524 147
AK297847.1 - 18 - 19 2713 147
AL136738.1 - 9 - 10 1318 147
AY766164.1 - 16 - 17 2413 147
BC025262.1 - 8 - 9 1336 147
CA310484.1 - 3 - 4 147
DQ768089.1 - 16 - 17 2337 147

View Full Product Details