The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018442.3 | NP_060912.2 | 10 - 11 | 1727 | 69 |
XM_005245332.4 | XP_005245389.1 | 10 - 11 | 1668 | 69 | |
XM_017001780.1 | XP_016857269.1 | 10 - 11 | 1476 | 69 | |
GenBank mRNA | AF124434.1 | - | 8 - 9 | 1165 | 69 |
AF150734.1 | - | 7 - 8 | 1027 | 69 | |
AL136738.1 | - | 2 - 3 | 312 | 69 |