Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PLEKHA6
Entrez Gene ID
22874
Gene Name
pleckstrin homology domain containing A6
Gene Aliases
PEPP-3, PEPP3
Chromosome Location
Chr.1: 204218851 - 204378376 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014935.4 NP_055750.2 12 - 13 2159 92
XM_005244966.4 XP_005245023.2 15 - 16 2248 92
XM_005244967.4 XP_005245024.2 12 - 13 2085 92
XM_005244968.4 XP_005245025.2 12 - 13 2025 92
XM_006711212.3 XP_006711275.1 15 - 16 2534 92
XM_006711213.3 XP_006711276.2 14 - 15 2254 92
XM_006711214.3 XP_006711277.2 15 - 16 2308 92
XM_006711215.3 XP_006711278.2 13 - 14 2173 92
XM_006711217.3 XP_006711280.2 15 - 16 2308 92
XM_006711219.3 XP_006711282.2 12 - 13 2086 92
XM_006711221.2 XP_006711284.1 15 - 16 2632 92
XM_006711222.3 XP_006711285.1 15 - 16 2617 92
XM_011509295.2 XP_011507597.1 15 - 16 2308 92
XM_011509296.2 XP_011507598.1 14 - 15 2227 92
XM_011509297.2 XP_011507599.1 17 - 18 3134 92
XM_011509298.2 XP_011507600.1 15 - 16 2417 92
XM_011509299.2 XP_011507601.2 15 - 16 2423 92
XM_011509301.2 XP_011507603.1 15 - 16 6313 92
XM_011509304.2 XP_011507606.1 15 - 16 2262 92
XM_017000687.1 XP_016856176.1 15 - 16 2308 92
XM_017000688.1 XP_016856177.1 14 - 15 2254 92
XM_017000689.1 XP_016856178.1 13 - 14 2113 92
XM_017000690.1 XP_016856179.1 15 - 16 6253 92
XM_017000691.1 XP_016856180.1 10 - 11 4706 92
XM_017000692.1 XP_016856181.1 10 - 11 4646 92
GenBank mRNA AB023186.1 - 12 - 13 2146 92
BC152475.1 - 12 - 13 2146 92

View Full Product Details