Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- PLEKHA6
- Entrez Gene ID
-
22874
- Gene Name
- pleckstrin homology domain containing A6
- Gene Aliases
-
PEPP-3, PEPP3
- Chromosome Location
-
Chr.1: 204218851 - 204378376 on Build GRCh38
View Full Product Details