The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018288.3 | NP_060758.2 | 8 - 9 | 1037 | 73 |
NM_133325.2 | NP_579866.2 | 8 - 9 | 1031 | 73 | |
GenBank mRNA | AJ420510.1 | - | 7 - 8 | 931 | 73 |
AK001837.1 | - | 6 - 7 | 701 | 73 | |
AK222612.1 | - | 6 - 7 | 696 | 73 | |
AK296349.1 | - | 4 - 5 | 464 | 73 | |
BC020954.2 | - | 6 - 7 | 728 | 73 | |
BC110323.1 | - | 8 - 9 | 1008 | 73 | |
CR457273.1 | - | 6 - 7 | 694 | 73 | |
KC839988.1 | - | 8 - 9 | 1017 | 73 | |
KC839989.1 | - | 8 - 9 | 1032 | 73 | |
KC839990.1 | - | 8 - 9 | 1017 | 73 |