Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PHF10
Entrez Gene ID
55274
Gene Name
PHD finger protein 10
Gene Aliases
BAF45A, XAP135
Chromosome Location
Chr.6: 169703902 - 169724049 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018288.3 NP_060758.2 1 - 2 164 76
NM_133325.2 NP_579866.2 1 - 2 164 76
XM_017010998.1 XP_016866487.1 1 - 2 203 76
GenBank mRNA AJ420510.1 - 1 - 2 168 76
BC110323.1 - 1 - 2 135 76
DB460742.1 - 1 - 2 164 76
KC839989.1 - 1 - 2 159 76

View Full Product Details