The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001029858.3 | NP_001025029.2 | 1 - 2 | 371 | 62 |
XM_005266865.4 | XP_005266922.1 | 1 - 2 | 671 | 62 | |
GenBank mRNA | BC028615.1 | - | 1 - 2 | 374 | 62 |
BC036493.1 | - | 1 - 2 | 140 | 61 | |
BC114925.1 | - | 2 - 3 | 245 | 62 |