Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC35F1
Entrez Gene ID
222553
Gene Name
solute carrier family 35 member F1
Gene Aliases
C6orf169, dJ230I3.1
Chromosome Location
Chr.6: 117907226 - 118317676 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029858.3 NP_001025029.2 4 - 5 834 73
XM_005266865.4 XP_005266922.1 4 - 5 1134 73
GenBank mRNA AK128717.1 - 2 - 3 182 73
AK295860.1 - 4 - 5 531 73
BC028615.1 - 4 - 5 837 73
BC036493.1 - 4 - 5 603 73
BC114925.1 - 5 - 6 708 73
BX640761.1 - 1 - 2 95 73

View Full Product Details