Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MAST2
Entrez Gene ID
23139
Gene Name
microtubule associated serine/threonine kinase 2
Gene Aliases
MAST205, MTSSK
Chromosome Location
Chr.1: 45786992 - 46036124 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001319245.1 NP_001306174.1 18 - 19 2498 84
NM_001324320.1 NP_001311249.1 19 - 20 2525 84
NM_001324321.1 NP_001311250.1 18 - 19 2611 84
NM_015112.2 NP_055927.2 18 - 19 2498 84
XM_005270655.3 XP_005270712.1 17 - 18 2218 84
XM_005270656.4 XP_005270713.1 17 - 18 2087 84
XM_006710477.3 XP_006710540.1 16 - 17 1895 84
XM_011541059.2 XP_011539361.1 20 - 21 2732 84
XM_011541061.2 XP_011539363.1 20 - 21 2732 84
XM_011541062.2 XP_011539364.1 19 - 20 2711 84
XM_011541063.1 XP_011539365.1 20 - 21 2296 84
XM_011541064.2 XP_011539366.1 20 - 21 2293 84
XM_011541065.1 XP_011539367.1 20 - 21 2395 84
XM_011541067.2 XP_011539369.1 16 - 17 2209 84
XM_011541068.1 XP_011539370.1 15 - 16 1826 84
XM_011541069.2 XP_011539371.1 20 - 21 2732 84
XM_017000752.1 XP_016856241.1 18 - 19 2725 84
XM_017000753.1 XP_016856242.1 17 - 18 2059 84
XM_017000754.1 XP_016856243.1 16 - 17 2038 84
XM_017000755.1 XP_016856244.1 17 - 18 2530 84
GenBank mRNA AB018350.2 - 7 - 8 810 84
AB047005.1 - 18 - 19 2498 84
AK027123.1 - 16 - 17 2037 84
AK096519.1 - 17 - 18 1822 84
BC006166.1 - 10 - 11 1166 84
BC015816.2 - 10 - 11 1166 84
BC065499.1 - 18 - 19 2472 84

View Full Product Details