Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MAST2
Entrez Gene ID
23139
Gene Name
microtubule associated serine/threonine kinase 2
Gene Aliases
MAST205, MTSSK
Chromosome Location
Chr.1: 45786992 - 46036124 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001319245.1 NP_001306174.1 24 - 25 3471 89
NM_001324320.1 NP_001311249.1 25 - 26 3498 89
NM_001324321.1 NP_001311250.1 24 - 25 3584 89
NM_015112.2 NP_055927.2 24 - 25 3471 89
XM_005270655.3 XP_005270712.1 23 - 24 3191 89
XM_005270656.4 XP_005270713.1 23 - 24 3060 89
XM_006710477.3 XP_006710540.1 22 - 23 2868 89
XM_011541059.2 XP_011539361.1 26 - 27 3705 89
XM_011541061.2 XP_011539363.1 26 - 27 3705 89
XM_011541062.2 XP_011539364.1 25 - 26 3684 89
XM_011541063.1 XP_011539365.1 26 - 27 3269 89
XM_011541064.2 XP_011539366.1 26 - 27 3266 89
XM_011541065.1 XP_011539367.1 26 - 27 3368 89
XM_011541067.2 XP_011539369.1 22 - 23 3182 89
XM_011541068.1 XP_011539370.1 21 - 22 2799 89
XM_011541069.2 XP_011539371.1 26 - 27 3705 89
XM_017000752.1 XP_016856241.1 24 - 25 3698 89
XM_017000753.1 XP_016856242.1 23 - 24 3032 89
XM_017000754.1 XP_016856243.1 22 - 23 3011 89
XM_017000755.1 XP_016856244.1 23 - 24 3503 89
GenBank mRNA AB018350.2 - 13 - 14 1783 89
AB047005.1 - 24 - 25 3471 89
AL833919.1 - 4 - 5 694 89
BC006166.1 - 16 - 17 2139 89
BC015816.2 - 16 - 17 2139 89
BC065499.1 - 24 - 25 3445 89

View Full Product Details