The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003647.2 | NP_003638.1 | 11 - 12 | 1703 | 63 |
XM_011525394.2 | XP_011523696.1 | 11 - 12 | 1681 | 63 | |
XM_017025243.1 | XP_016880732.1 | 11 - 12 | 2070 | 63 | |
XM_017025244.1 | XP_016880733.1 | 10 - 11 | 1612 | 63 | |
GenBank mRNA | AK293039.1 | - | 11 - 12 | 1650 | 63 |
BC130629.1 | - | 10 - 11 | 1547 | 63 | |
BC130631.1 | - | 10 - 11 | 1547 | 63 | |
U49379.1 | - | 11 - 12 | 1610 | 63 |