The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003647.2 | NP_003638.1 | 3 - 4 | 808 | 143 |
XM_011525394.2 | XP_011523696.1 | 3 - 4 | 786 | 143 | |
XM_011525395.2 | XP_011523697.1 | 3 - 4 | 793 | 143 | |
XM_011525396.2 | XP_011523698.1 | 3 - 4 | 795 | 143 | |
XM_017025243.1 | XP_016880732.1 | 3 - 4 | 803 | 143 | |
XM_017025244.1 | XP_016880733.1 | 3 - 4 | 789 | 143 | |
XM_017025245.1 | XP_016880734.1 | 3 - 4 | 792 | 143 | |
GenBank mRNA | AB209365.1 | - | 2 - 3 | 458 | 143 |
AK293039.1 | - | 3 - 4 | 755 | 143 | |
AK310668.1 | - | 3 - 4 | 776 | 143 | |
AL555194.3 | - | 3 - 4 | 807 | 143 | |
BC130629.1 | - | 2 - 3 | 652 | 143 | |
BC130631.1 | - | 2 - 3 | 652 | 143 | |
DB221284.1 | - | 2 - 3 | 470 | 143 | |
U49379.1 | - | 3 - 4 | 715 | 143 |