Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ASCC3
Entrez Gene ID
10973
Gene Name
activating signal cointegrator 1 complex subunit 3
Gene Aliases
ASC1p200, HELIC1, RNAH
Chromosome Location
Chr.6: 100508194 - 100881372 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006828.3 NP_006819.2 41 - 42 6816 95
XM_011535394.2 XP_011533696.1 41 - 42 6826 95
XM_011535395.2 XP_011533697.1 42 - 43 7051 95
XM_011535396.2 XP_011533698.1 40 - 41 6660 95
XM_017010205.1 XP_016865694.1 42 - 43 7072 95
XM_017010206.1 XP_016865695.1 42 - 43 6850 95
GenBank mRNA AL834463.1 - 40 - 41 6501 95
BC039857.1 - 5 - 6 736 95

View Full Product Details