Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ASCC3
Entrez Gene ID
10973
Gene Name
activating signal cointegrator 1 complex subunit 3
Gene Aliases
ASC1p200, HELIC1, RNAH
Chromosome Location
Chr.6: 100508194 - 100881372 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001284271.1 NP_001271200.1 7 - 8 1623 117
NM_006828.3 NP_006819.2 7 - 8 1623 117
XM_011535394.2 XP_011533696.1 7 - 8 1633 117
XM_011535395.2 XP_011533697.1 8 - 9 1858 117
XM_011535396.2 XP_011533698.1 6 - 7 1467 117
XM_017010205.1 XP_016865694.1 8 - 9 1879 117
XM_017010206.1 XP_016865695.1 7 - 8 1615 117
GenBank mRNA AK299117.1 - 6 - 7 1472 117
AY013288.1 - 3 - 4 442 117
BC026066.1 - 7 - 8 1620 117

View Full Product Details