Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WDR19
Entrez Gene ID
57728
Gene Name
WD repeat domain 19
Gene Aliases
ATD5, CED4, DYF-2, IFT144, NPHP13, ORF26, Oseg6, PWDMP, SRTD5
Chromosome Location
Chr.4: 39182404 - 39285810 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001317924.1 NP_001304853.1 5 - 6 558 99
NM_025132.3 NP_079408.3 6 - 7 674 99
XM_011513724.1 XP_011512026.1 6 - 7 584 99
XM_011513725.2 XP_011512027.1 5 - 6 539 99
XM_011513726.2 XP_011512028.1 5 - 6 447 99
XM_011513727.1 XP_011512029.1 4 - 5 367 99
XM_017008501.1 XP_016863990.1 4 - 5 367 99
GenBank mRNA AK294730.1 - 5 - 6 433 99
AK304161.1 - 6 - 7 547 99
AY029257.1 - 6 - 7 674 99
BC032578.1 - 5 - 6 553 99

View Full Product Details