The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001127487.1 | NP_001120959.1 | 38 - 39 | 6590 | 61 |
NM_001458.4 | NP_001449.3 | 39 - 40 | 6689 | 61 | |
GenBank mRNA | AB208865.1 | - | 16 - 17 | 2331 | 61 |
AB371585.1 | - | 39 - 40 | 6709 | 61 | |
AF089841.1 | - | 39 - 40 | 6529 | 61 | |
AF146692.1 | - | 38 - 39 | 6593 | 61 | |
AJ012737.1 | - | 38 - 39 | 6399 | 61 |