The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080527.1 | NP_001073996.1 | 17 - 18 | 2197 | 66 |
XM_006712539.3 | XP_006712602.1 | 17 - 18 | 2310 | 66 | |
XM_011511218.2 | XP_011509520.1 | 17 - 18 | 2310 | 66 | |
XM_017004169.1 | XP_016859658.1 | 17 - 18 | 2310 | 66 |