The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199763.1 | NP_001186692.1 | 1 - 2 | 356 | 111 |
NM_002846.3 | NP_002837.1 | 1 - 2 | 356 | 111 | |
XM_011511566.1 | XP_011509868.1 | 1 - 2 | 205 | 111 | |
XM_017004609.1 | XP_016860098.1 | 1 - 2 | 205 | 111 | |
XM_017004610.1 | XP_016860099.1 | 1 - 2 | 205 | 111 | |
XM_017004611.1 | XP_016860100.1 | 1 - 2 | 205 | 111 | |
XM_017004612.1 | XP_016860101.1 | 1 - 2 | 205 | 111 | |
GenBank mRNA | AK291469.1 | - | 1 - 2 | 281 | 111 |
AK294490.1 | - | 1 - 2 | 135 | 111 | |
AK314574.1 | - | 1 - 2 | 120 | 111 | |
BC070053.1 | - | 1 - 2 | 182 | 111 | |
L18983.1 | - | 1 - 2 | 188 | 111 |