Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PTPRN
Entrez Gene ID
5798
Gene Name
protein tyrosine phosphatase, receptor type N
Gene Aliases
IA-2, IA-2/PTP, IA2, ICA512, R-PTP-N
Chromosome Location
Chr.2: 219289623 - 219309573 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199763.1 NP_001186692.1 2 - 3 408 56
NM_001199764.1 NP_001186693.1 2 - 3 178 56
NM_002846.3 NP_002837.1 2 - 3 408 56
XM_011511566.1 XP_011509868.1 2 - 3 257 56
XM_017004609.1 XP_016860098.1 2 - 3 257 56
XM_017004610.1 XP_016860099.1 2 - 3 257 56
XM_017004611.1 XP_016860100.1 2 - 3 257 56
XM_017004612.1 XP_016860101.1 2 - 3 257 56
GenBank mRNA AK291469.1 - 2 - 3 333 56
AK294490.1 - 2 - 3 187 56
AK296335.1 - 2 - 3 155 56
AK314574.1 - 2 - 3 172 56
BC070053.1 - 2 - 3 234 56
DC352946.1 - 2 - 3 178 56
L18983.1 - 2 - 3 240 56

View Full Product Details