The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032526.2 | NP_115915.1 | 2 - 3 | 302 | 79 |
GenBank mRNA | AF331801.1 | - | 2 - 3 | 302 | 79 |
AY028778.1 | - | 2 - 3 | 293 | 79 | |
BC103879.1 | - | 2 - 3 | 302 | 79 | |
BC103880.1 | - | 2 - 3 | 302 | 79 |