The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016141.3 | NP_057225.2 | 4 - 5 | 673 | 126 |
GenBank mRNA | AF078849.1 | - | 4 - 5 | 649 | 126 |
AK001081.1 | - | 1 - 2 | 169 | 126 | |
AK074348.1 | - | 4 - 5 | 657 | 126 | |
AK222573.1 | - | 4 - 5 | 126 | ||
AK222653.1 | - | 4 - 5 | 609 | 126 | |
BC131620.1 | - | 4 - 5 | 619 | 126 | |
BX649189.1 | - | 4 - 5 | 492 | 126 |