Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TYR
Entrez Gene ID
7299
Gene Name
tyrosinase
Gene Aliases
ATN, CMM8, OCA1, OCA1A, OCAIA, SHEP3
Chromosome Location
Chr.11: 89177565 - 89295759 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000372.4 NP_000363.1 3 - 4 1266 107
XM_011542970.2 XP_011541272.1 3 - 4 1573 107
GenBank mRNA AB775899.1 - 3 - 4 1184 107
AB775900.1 - 3 - 4 1184 107
AB775901.1 - 3 - 4 1184 107
J03581.1 - 3 - 4 1178 107
M27160.1 - 3 - 4 1686 107
M74314.1 - 3 - 4 1224 107
U01873.1 - 3 - 4 1184 107
Y00819.1 - 3 - 4 1190 107

View Full Product Details