Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RGS6
Entrez Gene ID
9628
Gene Name
regulator of G-protein signaling 6
Gene Aliases
GAP
Chromosome Location
Chr.14: 71932100 - 72632478 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001204416.2 NP_001191345.1 2 - 3 361 85
NM_001204417.2 NP_001191346.1 2 - 3 361 85
NM_001204418.2 NP_001191347.1 2 - 3 361 85
NM_001204419.2 NP_001191348.1 2 - 3 361 85
NM_001204420.2 NP_001191349.1 2 - 3 361 85
NM_001204421.2 NP_001191350.1 2 - 3 361 85
NM_001204422.2 NP_001191351.1 2 - 3 361 85
NM_001204424.1 NP_001191353.1 2 - 3 599 85
NM_004296.6 NP_004287.3 2 - 3 361 85
NR_135235.1 2 - 3 361 85
XM_017021818.1 XP_016877307.1 2 - 3 261 85
XM_017021819.1 XP_016877308.1 2 - 3 261 85
XM_017021820.1 XP_016877309.1 1 - 2 96 85
XM_017021821.1 XP_016877310.1 1 - 2 96 85
XM_017021822.1 XP_016877311.1 1 - 2 96 85
XM_017021823.1 XP_016877312.1 1 - 2 96 85
XM_017021824.1 XP_016877313.1 2 - 3 261 85
XM_017021825.1 XP_016877314.1 1 - 2 96 85
XM_017021826.1 XP_016877315.1 1 - 2 96 85
XM_017021827.1 XP_016877316.1 1 - 2 96 85
XM_017021828.1 XP_016877317.1 1 - 2 96 85
XM_017021829.1 XP_016877318.1 2 - 3 261 85
XM_017021830.1 XP_016877319.1 1 - 2 96 85
XM_017021831.1 XP_016877320.1 1 - 2 96 85
XM_017021832.1 XP_016877321.1 1 - 2 96 85
XM_017021833.1 XP_016877322.1 1 - 2 96 85
GenBank mRNA AB209462.1 - 2 - 3 167 85
AF073920.2 - 2 - 3 283 85
AF107619.1 - 1 - 2 76 85
AF107620.1 - 1 - 2 76 85
AF156932.1 - 1 - 2 76 85
AK294486.1 - 2 - 3 599 85
AK313481.1 - 2 - 3 421 85
AY309097.1 - 1 - 2 76 85
AY309098.1 - 1 - 2 76 85
AY309099.1 - 1 - 2 76 85
AY309100.1 - 1 - 2 76 85
BC104877.1 - 2 - 3 254 85
BC113572.1 - 2 - 3 254 85
BC143699.1 - 2 - 3 254 85
DC317315.1 - 2 - 3 361 85
DC343081.1 - - 330 85

View Full Product Details