Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RGS6
Entrez Gene ID
9628
Gene Name
regulator of G-protein signaling 6
Gene Aliases
GAP
Chromosome Location
Chr.14: 71932100 - 72632478 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001204416.2 NP_001191345.1 5 - 6 624 83
NM_001204417.2 NP_001191346.1 5 - 6 624 83
NM_001204418.2 NP_001191347.1 5 - 6 624 83
NM_001204419.2 NP_001191348.1 5 - 6 624 83
NM_001204420.2 NP_001191349.1 5 - 6 624 83
NM_001204421.2 NP_001191350.1 5 - 6 624 83
NM_001204422.2 NP_001191351.1 5 - 6 624 83
NM_001204423.1 NP_001191352.1 4 - 5 389 83
NM_001204424.1 NP_001191353.1 5 - 6 862 83
NM_004296.6 NP_004287.3 5 - 6 624 83
NR_135235.1 5 - 6 624 83
XM_011537393.2 XP_011535695.1 4 - 5 873 83
XM_011537397.1 XP_011535699.1 2 - 3 155 83
XM_011537407.2 XP_011535709.1 4 - 5 876 83
XM_017021818.1 XP_016877307.1 5 - 6 524 83
XM_017021819.1 XP_016877308.1 5 - 6 524 83
XM_017021820.1 XP_016877309.1 4 - 5 359 83
XM_017021821.1 XP_016877310.1 4 - 5 359 83
XM_017021822.1 XP_016877311.1 4 - 5 359 83
XM_017021823.1 XP_016877312.1 4 - 5 359 83
XM_017021824.1 XP_016877313.1 5 - 6 524 83
XM_017021825.1 XP_016877314.1 4 - 5 359 83
XM_017021826.1 XP_016877315.1 4 - 5 359 83
XM_017021827.1 XP_016877316.1 4 - 5 359 83
XM_017021828.1 XP_016877317.1 4 - 5 359 83
XM_017021829.1 XP_016877318.1 5 - 6 524 83
XM_017021830.1 XP_016877319.1 4 - 5 359 83
XM_017021831.1 XP_016877320.1 4 - 5 359 83
XM_017021832.1 XP_016877321.1 4 - 5 359 83
XM_017021833.1 XP_016877322.1 4 - 5 359 83
XM_017021834.1 XP_016877323.1 2 - 3 155 83
GenBank mRNA AB209462.1 - 5 - 6 430 83
AF073920.2 - 5 - 6 546 83
AF107619.1 - 4 - 5 339 83
AF107620.1 - 4 - 5 339 83
AF156932.1 - 4 - 5 339 83
AK294486.1 - 5 - 6 862 83
AK294895.1 - 4 - 5 389 83
AK302319.1 - 3 - 4 235 83
AK310463.1 - 4 - 5 498 83
AK313481.1 - 5 - 6 684 83
AY309097.1 - 4 - 5 339 83
AY309098.1 - 4 - 5 339 83
AY309099.1 - 4 - 5 339 83
AY309100.1 - 4 - 5 339 83
BC104877.1 - 5 - 6 517 83
BC113572.1 - 5 - 6 517 83
BC143699.1 - 5 - 6 517 83
DA386861.1 - 3 - 4 198 83

View Full Product Details