Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SPOPL
Entrez Gene ID
339745
Gene Name
speckle type BTB/POZ protein like
Gene Aliases
BTBD33
Chromosome Location
Chr.2: 138501780 - 138573235 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001001664.2 NP_001001664.1 8 - 9 1236 70
XM_005263655.4 XP_005263712.1 9 - 10 1199 70
XM_005263656.4 XP_005263713.1 9 - 10 1128 70
XM_005263657.4 XP_005263714.1 8 - 9 1041 70
GenBank mRNA AK293850.1 - 2 - 3 151 70
BC071613.1 - 8 - 9 1215 70

View Full Product Details