The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173462.3 | NP_775733.3 | 17 - 18 | 2270 | 102 |
XM_011537290.2 | XP_011535592.1 | 18 - 19 | 2351 | 102 | |
XM_011537291.2 | XP_011535593.1 | 19 - 20 | 2398 | 102 | |
XM_011537292.2 | XP_011535594.1 | 18 - 19 | 2274 | 102 | |
XM_011537293.2 | XP_011535595.1 | 17 - 18 | 2270 | 102 | |
XM_011537294.2 | XP_011535596.1 | 17 - 18 | 2275 | 102 | |
XM_011537295.2 | XP_011535597.1 | 17 - 18 | 2270 | 102 | |
XM_011537296.2 | XP_011535598.1 | 17 - 18 | 2270 | 102 | |
XM_017021749.1 | XP_016877238.1 | 17 - 18 | 2270 | 102 | |
XM_017021750.1 | XP_016877239.1 | 17 - 18 | 2273 | 102 | |
GenBank mRNA | AK127354.1 | - | 3 - 4 | 419 | 102 |
AK131073.1 | - | 10 - 11 | 1563 | 102 | |
AK293773.1 | - | 17 - 18 | 2412 | 102 | |
BC144315.1 | - | 16 - 17 | 2188 | 102 | |
BX537757.1 | - | 3 - 4 | 396 | 102 |