The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173462.3 | NP_775733.3 | 8 - 9 | 864 | 147 |
XM_011537290.2 | XP_011535592.1 | 9 - 10 | 945 | 147 | |
XM_011537291.2 | XP_011535593.1 | 10 - 11 | 992 | 147 | |
XM_011537292.2 | XP_011535594.1 | 9 - 10 | 868 | 147 | |
XM_011537293.2 | XP_011535595.1 | 8 - 9 | 864 | 147 | |
XM_011537294.2 | XP_011535596.1 | 8 - 9 | 869 | 147 | |
XM_011537295.2 | XP_011535597.1 | 8 - 9 | 864 | 147 | |
XM_011537296.2 | XP_011535598.1 | 8 - 9 | 864 | 147 | |
XM_017021749.1 | XP_016877238.1 | 8 - 9 | 864 | 147 | |
XM_017021750.1 | XP_016877239.1 | 8 - 9 | 867 | 147 | |
GenBank mRNA | AK131073.1 | - | 1 - 2 | 157 | 147 |
AK293773.1 | - | 8 - 9 | 1006 | 147 | |
AK294560.1 | - | 8 - 9 | 997 | 147 | |
BC136623.1 | - | 8 - 9 | 863 | 147 | |
BC144315.1 | - | 7 - 8 | 782 | 147 | |
BX470414.1 | - | 4 - 5 | 147 |