The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012216.3 | NP_036348.2 | 1 - 2 | 574 | 82 |
NM_052817.2 | NP_438112.2 | 1 - 2 | 574 | 82 | |
GenBank mRNA | Y18880.1 | - | 1 - 2 | 198 | 82 |