The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012216.3 | NP_036348.2 | 7 - 8 | 2011 | 104 |
XM_005262062.4 | XP_005262119.1 | 7 - 8 | 6274 | 104 | |
GenBank mRNA | Y18880.1 | - | 7 - 8 | 1635 | 104 |