The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012216.3 | NP_036348.2 | 8 - 9 | 2171 | 72 |
NM_052817.2 | NP_438112.2 | 8 - 9 | 2081 | 72 | |
XM_005262062.4 | XP_005262119.1 | 8 - 9 | 6434 | 72 | |
XM_017029239.1 | XP_016884728.1 | 8 - 9 | 6344 | 72 | |
GenBank mRNA | AF196481.1 | - | 8 - 9 | 1521 | 72 |
AY625004.1 | - | 7 - 8 | 1448 | 72 | |
BC017707.1 | - | 7 - 8 | 1475 | 72 | |
BT006663.1 | - | 7 - 8 | 1448 | 72 | |
Y18880.1 | - | 8 - 9 | 1795 | 72 |