The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001326578.1 | NP_001313507.1 | 7 - 8 | 862 | 149 |
NM_001326579.1 | NP_001313508.1 | 7 - 8 | 649 | 149 | |
NM_001326580.1 | NP_001313509.1 | 7 - 8 | 785 | 149 | |
NM_003569.2 | NP_003560.2 | 7 - 8 | 785 | 149 | |
XM_011536178.1 | XP_011534480.1 | 7 - 8 | 701 | 149 | |
GenBank mRNA | AK294911.1 | - | 7 - 8 | 534 | 149 |
BC011975.1 | - | 7 - 8 | 649 | 149 | |
U77942.1 | - | 7 - 8 | 614 | 149 |